3,7-Dibromodibenzothiophene 5,5-Dioxide ≥97.0% (by GC)
Catalog # TCIAD5052-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:374.05 g/mol
Melting Pt:312°C
CAS Number:83834-12-2
Formula:C₁₂H₆Br₂O₂S
MDL Number:MFCD00227704