Order Entry
Italy
ContactUsLinkComponent
Dimethyl (1-diazo-2-oxopropyl)phosphonate 10% in acetonitrile
Flammable
Toxic
Health
Dimethyl (1-diazo-2-oxopropyl)phosphonate 10% in acetonitrile
Supplier:  TCI
Certificates
Flammable
Toxic
Health
Dimethyl (1-diazo-2-oxopropyl)phosphonate 10% in acetonitrile
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Concentration (HNMR):
    10.0 to 11.0 w/w%

Specification Test Results

Frequently Bought Together