Order Entry
Italy
ContactUsLinkComponent
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98.0% (by HPLC, titration analysis)
Irritant
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    146.0 to 150.0 deg-C

Specification Test Results

Frequently Bought Together