Order Entry
Italy
ContactUsLinkComponent
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98.0% (by GC)
Irritant
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    186.0 to 190.0 deg-C

Specification Test Results

Frequently Bought Together