2,8-Dibromodibenzothiophene 5,5-Dioxide ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:40307-15-1
:
Formula:C₁₂H₆Br₂O₂S
MDL Number:MFCD00093102
:374.05 g/mol
Melting Pt:361°C
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together