Order Entry
Italy
ContactUsLinkComponent
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione ≥98.0% (by HPLC, total nitrogen)
Irritant
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
Irritant
5,6-Diamino-1-methylpyrimidine-2,4(1H,3H)-dione ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Frequently Bought Together