Order Entry
Italy
ContactUsLinkComponent
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥98.0% (by HPLC)
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Optical purity (LC):
    min. 98.0 ee%
  • Elemental analysis (Nitrogen):
    2.20 to 3.00 %

Specification Test Results

Frequently Bought Together