(11bR)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥98.0% (by HPLC)
: TCI
:415918-91-1
:MFCD08561138
:C₃₆H₃₀NO₂P
:539.61 g/mol
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 95.0 area%
- Optical purity (LC):min. 98.0 ee%
- Elemental analysis (Nitrogen):2.20 to 3.00 %
Frequently Bought Together