
Dimethyl acetal for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:534-15-6
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00008493
:90.12 g/mol
Merck Index:13,03253
Boiling Pt:64 °C (1013 hPa)
Melting Pt:–113 °C
Density:0.8505 g/cm³ (20 °C)
UN:2377
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:208-589-8
:GHS02
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-113 °C
- Molar Mass:90.12 g/mol
- Boiling Point:63 - 65 °C (1013 hPa)
- Vapor Pressure:200 hPa (20 °C)
- Flash Point:-17 °C
- Refractive Index:1.3665 (20 °C, 589 nm)
- Density:0.85 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Frequently Bought Together