Order Entry
Italy
ContactUsLinkComponent
1H,1H,7H-Dodecafluoroheptyl acrylate ≥97.0% (by GC) stabilised
Irritant
1H,1H,7H-Dodecafluoroheptyl acrylate ≥97.0% (by GC) stabilised
Catalog # TCIAD4989-5G
Supplier:  TCI
CAS Number:  
Irritant
1H,1H,7H-Dodecafluoroheptyl acrylate ≥97.0% (by GC) stabilised
Catalog # TCIAD4989-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :386.14 g/mol
Boiling Pt:107°C
Flash Pt:102°C
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
EINECS:221-064-8
 :GHS07
Storage Temperature:Refrigerator
CAS Number:2993-85-3
Formula:C₁₀H₆F₁₂O₂
MDL Number:MFCD00080746