
Dithieno[3,2-c:2',3'-e]oxepine-4,6-dione ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1043023-52-4
:
Formula:C₁₀H₄O₃S₂
:236.26 g/mol
Melting Pt:263°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Methanolysis Method):min. 98.0 %
Specification Test Results
Frequently Bought Together