Order Entry
Italy
ContactUsLinkComponent
N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
N,N'-Di(4-biphenylyl)-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    211.0 to 215.0 deg-C

Specification Test Results

Frequently Bought Together