1,4-Di(1-pyrenyl)benzene ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:475460-77-6
:MFCD27923028
Formula:C₃₈H₂₂
MDL Number:MFCD12022457
:478.59 g/mol
Specification Test Results
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together