Order Entry
Italy
ContactUsLinkComponent
 
Corrosive
Irritant
Health
2,4-Dichlorophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAD4912-5G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Health
2,4-Dichlorophenyl isothiocyanate ≥98.0% (by GC)
Catalog # TCIAD4912-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    40.0 to 44.0 deg-C

Specification Test Results

About this item

 :
ADR:8,III
 :204.08 g/mol
EINECS:229-527-6
UN:1759
 :GHS05
 :GHS07
 :GHS08
CAS Number:6590-96-1
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00037827