
5,6-Dichloropyridin-3-ol ≥98.0% (by GC)
Catalog # TCIAD4899-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:184.0 to 188.0 deg-C
Specification Test Results
About this item
:
:163.9900 g/mol
:GHS07
CAS Number:110860-92-9
Formula:C₅H₃Cl₂NO
MDL Number:MFCD09835219