Order Entry
Italy
ContactUsLinkComponent
2,4-Dichloro-1,8-naphthyridine ≥96.0% (by GC, titration analysis)
Irritant
2,4-Dichloro-1,8-naphthyridine ≥96.0% (by GC, titration analysis)
Catalog # TCIAD4896-1G
Supplier:  TCI
CAS Number:  
Irritant
2,4-Dichloro-1,8-naphthyridine ≥96.0% (by GC, titration analysis)
Catalog # TCIAD4896-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Melting point:
    123.0 to 127.0 deg-C

Specification Test Results

About this item

 :
 :199.03 g/mol
 :GHS07
CAS Number:59514-89-5
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00234380