Order Entry
Italy
ContactUsLinkComponent
5,5'''-Didecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by HPLC)
5,5'''-Didecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by HPLC)
Catalog # TCIAD4888-100MG
Supplier:  TCI
CAS Number:  
5,5'''-Didecyl-2,2':5',2'':5'',2'''-quaterthiophene ≥95.0% (by HPLC)
Catalog # TCIAD4888-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :611.04 g/mol
CAS Number:514188-77-3
Formula:C₃₆H₅₀S₄