1,5-Diethynyl-2,4-dimethylbenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1379822-09-9
:
Formula:C₁₂H₁₀
:154.21 g/mol
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:56.0 to 60.0 deg-C
Specification Test Results
Frequently Bought Together