
2,4-Dichlorobenzhydrazide ≥98.0% (by GC, titration analysis)
: TCI
:5814-06-2
:
:C₇H₆Cl₂N₂O
:MFCD00025113
:205.04 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:167.0 to 171.0 deg-C
Frequently Bought Together