Order Entry
Italy
ContactUsLinkComponent
N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine ≥98.0% (by HPLC, total nitrogen)
N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
N,N'-Di-1-naphthyl-N,N'-di-2-naphthylbenzidine ≥98.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Frequently Bought Together