Order Entry
Italy
ContactUsLinkComponent
5,6-Diamino-1,3-dipropyluracil hydrochloride ≥98.0% (by HPLC)
Irritant
5,6-Diamino-1,3-dipropyluracil hydrochloride ≥98.0% (by HPLC)
Catalog # TCIAD4810-1G
Supplier:  TCI
CAS Number:  
Irritant
5,6-Diamino-1,3-dipropyluracil hydrochloride ≥98.0% (by HPLC)
Catalog # TCIAD4810-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    242.0 to 246.0 deg-C

Specification Test Results

About this item

 :
 :262.74 g/mol
 :GHS07
CAS Number:324002-49-5
Formula:C₁₀H₁₈N₄O₂·HCl
MDL Number:MFCD06797515