Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromo-4,4-di-n-octyldithieno[3,2-b:2',3'-d]silole ≥98.0% (by GC)
2,6-Dibromo-4,4-di-n-octyldithieno[3,2-b:2',3'-d]silole ≥98.0% (by GC)
Catalog # TCIAD4799-100MG
Supplier:  TCI
CAS Number:  
2,6-Dibromo-4,4-di-n-octyldithieno[3,2-b:2',3'-d]silole ≥98.0% (by GC)
Catalog # TCIAD4799-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :576.57 g/mol
CAS Number:1160106-14-8
Formula:C₂₄H₃₆Br₂S₂Si
MDL Number:MFCD26793508