
1,3-Dibromo-5-hexylbenzene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:75894-97-2
:
Formula:C₁₂H₁₆Br₂
:320.07 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together