
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4462-55-9
:
Formula:C₁₁H₆Cl₃NO₂
MDL Number:MFCD00052556
EINECS:224-723-8
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:93.0 to 97.0 deg-C
Specification Test Results
Frequently Bought Together