
2,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7657-09-2
:
Formula:C₇H₃Br₂F₃
MDL Number:MFCD00013554
:303.9 g/mol
Boiling Pt:207°C
EINECS:231-620-1
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C
Specification Test Results
Frequently Bought Together