Order Entry
Italy
ContactUsLinkComponent
L-Aspartic aciddiethyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
L-Aspartic aciddiethyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
L-Aspartic aciddiethyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    105.0 to 109.0 deg-C
  • Specific rotation [a]20/D:
    +7.0 to +9.0 deg (C=1, H2O)

Specification Test Results

Frequently Bought Together