Order Entry
Italy
ContactUsLinkComponent
5,7-Dihydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by titrimetric analysis)
Irritant
5,7-Dihydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by titrimetric analysis)
Catalog # TCIAD4747-1G
Supplier:  TCI
CAS Number:  
Irritant
5,7-Dihydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by titrimetric analysis)
Catalog # TCIAD4747-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :192.17 g/mol
Melting Pt:280°C
EINECS:218-289-9
 :GHS07
CAS Number:2107-76-8
Formula:C₁₀H₈O₄
MDL Number:MFCD00016966