
2,4-Difluoro-6-nitroaniline ≥98.0% (by GC)
TCIAD4733-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 86.0 deg-C
:
:174.11 g/mol
:GHS07
:364-30-7
:C₆H₄F₂N₂O₂
:MFCD00034058