
3,6-Dibromophthalic anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:25834-16-6
:
Formula:C₈H₂Br₂O₃
MDL Number:MFCD01117424
:305.91 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Methanolysis Method):min. 98.0 %
- Melting point:207.0 to 212.0 deg-C
Specification Test Results
Frequently Bought Together