(S)-4,4'-Dibromo-1,1'-spirobiindane-7,7'-diol ≥98.0% (by GC)
TCIAD4718-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:159.0 to 163.0 deg-C
:
:410.11 g/mol
:636601-27-9
:C₁₇H₁₄Br₂O₂
:MFCD16660947