Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromo-4-n-octyldithieno[3,2-b:2',3'-d]pyrrole ≥94.0% (by GC)
2,6-Dibromo-4-n-octyldithieno[3,2-b:2',3'-d]pyrrole ≥94.0% (by GC)
Catalog # TCIAD4714-200MG
Supplier:  TCI
CAS Number:  
2,6-Dibromo-4-n-octyldithieno[3,2-b:2',3'-d]pyrrole ≥94.0% (by GC)
Catalog # TCIAD4714-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 94.0 %

Specification Test Results

About this item

 :
 :449.26 g/mol
CAS Number:1346688-54-7
Formula:C₁₆H₁₉Br₂NS₂