Order Entry
Italy
ContactUsLinkComponent
16-Dehydropregnenolone Acetate ≥98.0% (by HPLC)
16-Dehydropregnenolone Acetate ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
16-Dehydropregnenolone Acetate ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    172.0 to 176.0 deg-C
  • Specific rotation [a]20/D:
    -38.0 to -42.0 deg (C=1,CHCl3)

Specification Test Results

Related Information

Frequently Bought Together