9,9-Di-n-octylfluorene-2,7-dicarboxaldehyde ≥98.0% (by GC)
Catalog # TCIAD4671-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C
Specification Test Results
About this item
:
:446.68 g/mol
CAS Number:380600-91-9
Formula:C₃₁H₄₂O₂