Order Entry
Italy
ContactUsLinkComponent
 
1,5-Dibromo-2,6-dihydroxynaphthalene ≥98.0% (by GC)
 :  TCI
 
1,5-Dibromo-2,6-dihydroxynaphthalene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Brown to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together