Order Entry
Italy
ContactUsLinkComponent
N-[(11bS)-4,5-Dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepin-2-yl]trifluoromethanesulphonamide ≥90.0% (by HPLC)
Irritant
N-[(11bS)-4,5-Dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepin-2-yl]trifluoromethanesulphonamide ≥90.0% (by HPLC)
Catalog # TCIAD4663-20MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
N-[(11bS)-4,5-Dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepin-2-yl]trifluoromethanesulphonamide ≥90.0% (by HPLC)
Catalog # TCIAD4663-20MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    20 mg
  • Seq:
    2305

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 90.0 area%

Specification Test Results

About this item

 :
 :442.46 g/mol
 :GHS07
CAS Number:871915-77-4
Formula:C₂₃H₁₇F₃N₂O₂S