Order Entry
Italy
ContactUsLinkComponent
5,6-Diamino-1,3-diethyluracil hydrochloride ≥98.0% (by HPLC, titration analysis)
Irritant
5,6-Diamino-1,3-diethyluracil hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
5,6-Diamino-1,3-diethyluracil hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    235.0 to 239.0 deg-C

Specification Test Results

Frequently Bought Together