
1,8-Dibromonaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17135-74-9
:
Formula:C₁₀H₆Br₂
MDL Number:MFCD00183574
:285.97 g/mol
Boiling Pt:140°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
Specification Test Results
Frequently Bought Together