Order Entry
Italy
ContactUsLinkComponent
4,5-Diazafluorene-9-one O-(p-Toluenesulphonyl)oxime ≥98.0% (by HPLC)
Irritant
4,5-Diazafluorene-9-one O-(p-Toluenesulphonyl)oxime ≥98.0% (by HPLC)
Codice catalogo TCIAD4636-100MG
Supplier:  TCI
CAS Number:  
Irritant
4,5-Diazafluorene-9-one O-(p-Toluenesulphonyl)oxime ≥98.0% (by HPLC)
Codice catalogo TCIAD4636-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    11.40 to 12.50 %

Specification Test Results

About this item

 :
 :351.38 g/mol
 :GHS07
CAS Number:1655490-79-1
Formula:C₁₈H₁₃N₃O₃S