
Dimethyl [2,2'-bipyridine]-4,4'-dicarboxylate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:71071-46-0
:
Formula:C₁₄H₁₂N₂O₄
MDL Number:MFCD02669916
:272.2600 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:208.0 to 212.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together