Order Entry
Italy
ContactUsLinkComponent
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Irritant
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    192.0 to 196.0 deg-C

Specification Test Results

Frequently Bought Together