Order Entry
Italy
ContactUsLinkComponent
6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c]-1,2,4-triazolium tetrafluoroborate ≥98.0% (by HPLC)
Corrosive
6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c]-1,2,4-triazolium tetrafluoroborate ≥98.0% (by HPLC)
Catalog # TCIAD4624-200MG
Supplier:  TCI
CAS Number:  
Corrosive
6,7-Dihydro-2-pentafluorophenyl-5H-pyrrolo[2,1-c]-1,2,4-triazolium tetrafluoroborate ≥98.0% (by HPLC)
Catalog # TCIAD4624-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    11.00 to 12.20 %
  • Melting point:
    244.0 to 250.0 deg-C

Specification Test Results

About this item

 :
ADR:8,II
 :362.99 g/mol
UN:1759
 :GHS05
CAS Number:862095-91-8
Formula:C₁₁H₇BF₉N₃
MDL Number:MFCD08459335