4,7-Dibromo-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole ≥98.0% (by HPLC)
Catalog # TCIAD4622-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):4.00 to 4.50 %
- Melting point:59.0 to 63.0 deg-C
Specification Test Results
About this item
:
:662.61 g/mol
CAS Number:1313876-00-4
Formula:C₃₀H₅₀Br₂N₂O₂S
MDL Number:MFCD27923049