
1,3-Difluoro-5-iodobenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2265-91-0
:
Formula:C₆H₃F₂I
MDL Number:MFCD03094155
:239.99 g/mol
Boiling Pt:60°C
Flash Pt:82°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together