2,8-Dimethylanthra[2,3-b:6,7-b']dithiophene, purified by sublimation
TCIAD4617-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Elemental analysis (Carbon):71.50 to 79.00 %
- Elemental analysis (Sulfuride):19.50 to 21.00 %
:
:318.45 g/mol
:1019983-99-3
:C₂₀H₁₄S₂