Order Entry
Italy
ContactUsLinkComponent
 
(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial ≥98.0% (by GC)
  TCIAD4584-25G
 :  TCI
 :  
(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial ≥98.0% (by GC)
  TCIAD4584-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    164.0 to 168.0 deg-C

 

 

 :
 :164.20 g/mol
 :5056-17-7
 :C₁₀H₁₂O₂
 :MFCD11113099