
(R)-1-(2,6-Dichloro-3-fluorophenyl)ethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:330156-50-8
:
Formula:C₈H₇Cl₂FO
MDL Number:MFCD09863794
:209.0500 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:41.0 to 45.0 deg-C
- Specific rotation [a]20/D:-7.0 to -9.0 deg (C=5,MeOH)
Specification Test Results
Frequently Bought Together