
2,8-Dibromoindeno[1,2-b]fluorene-6,12-dione ≥98.0% (by titrimetric analysis)
TCIAD4573-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
:
:440.09 g/mol
:GHS07
:853234-57-8
:C₂₀H₈Br₂O₂
:MFCD22571712