
2,7-Dibromo-9,9-dipropylfluorene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:157771-56-7
:
Formula:C₁₉H₂₀Br₂
MDL Number:MFCD12024280
:408.18 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:136.0 to 140.0 deg-C
Specification Test Results
Frequently Bought Together