Order Entry
Italy
ContactUsLinkComponent
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥99.0% (by HPLC)
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥99.0% (by HPLC)
Catalog # TCIAD4568-200MG
Supplier:  TCI
CAS Number:  
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene ≥99.0% (by HPLC)
Catalog # TCIAD4568-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Yellow to Brown to Dark green clear liquid
  • Purity (HPLC):
    min. 99.0 area%

Specification Test Results

About this item

 :
 :560.49 g/mol
CAS Number:365547-21-3
Formula:C₂₅H₃₆Br₂S₂
MDL Number:MFCD16619302