Order Entry
Italy
ContactUsLinkComponent
4,4-Di-n-octyldithieno[3,2-b:2,3-d']silole ≥95.0% (by HPLC)
4,4-Di-n-octyldithieno[3,2-b:2,3-d']silole ≥95.0% (by HPLC)
Catalog # TCIAD4564-200MG
Supplier:  TCI
CAS Number:  
4,4-Di-n-octyldithieno[3,2-b:2,3-d']silole ≥95.0% (by HPLC)
Catalog # TCIAD4564-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Yellow to Orange to Green clear liquid
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :418.77 g/mol
CAS Number:1160106-12-6
Formula:C₂₄H₃₈S₂Si
MDL Number:MFCD26793507